Coolers for Reagent Reservoirs
Supplier: DIVERSIFIED BIOTECH
Certificates
About this item
These benchtop coolers are placed under the reservoirs and keep reagents at ~5 °C for approximately 3 hours.
- No need for a cumbersome ice bath, the frozen non toxic gel is contained in a sealed molded cavity
- Extends safe working time for temperature sensitive reagents
Frequently Bought Together