Screw caps for moulded pathology media vials, WHEATON®
Supplier: DWK Life Sciences
Certificates
About this item
Self-sealing PP caps provide leak proof seal, aluminium caps fitted with non-inhibitory rubber liners are suitable for storage of cultures and allow cell growth.
Frequently Bought Together