Industrial gloves, AlphaTec® 23-202
: Ansell
These PVC coated gloves with inner polar acrylic seamless lining provide chemical and liquid protection and good dexterity at low temperatures.
- Roughened sandblast finish on the glove and smooth finish on the gauntlet cuff
- Designed to protect at temperatures down to −30 °C
- Immediate heat feeling when wearing the glove
- Good grip in handling both slippery and wet materials
- Soft integral PVC coating offers chemical resistance
Ideal for outdoor applications including chemical handlings.
Gauge: 13
Colour: Blue
EN 388 (4121), EN 374, EN 421, EN 511 (021); Cat. III; AQL 1,5 Food contact approved
: EN 388:2016 (3121B), EN ISO 374-1 Type B (KLT), EN ISO 374-5 VIRUS, EN 421, EN 511 (021); Cat. III
Food contact approved
Food contact approved
- Catalog No:
- ANSE23-202/10
- ANSE23-202/9
- ANSE23-202/8
- Model:
- 23-202
- 23-202
- 23-202
- Ambidextrous / hand-specific:
- Hand-specific
- Hand-specific
- Hand-specific
- Disposable / reusable:
- Reusable
- Reusable
- Reusable
- Liner material:
- Acrylic
- Acrylic
- Acrylic
- Finish:
- Sandblast
- Sandblast
- Sandblast
- Cuff style:
- Gauntlet
- Gauntlet
- Gauntlet
- EN norm:
- EN 388:2016 (3121B), EN ISO 374-1 Type B (KLT), EN ISO 374-5 VIRUS, EN 421, EN 511 (021)
- EN 388:2016 (3121B), EN ISO 374-1 Type B (KLT), EN ISO 374-5 VIRUS, EN 421, EN 511 (021)
- EN 388:2016 (3121B), EN ISO 374-1 Type B (KLT), EN ISO 374-5 VIRUS, EN 421, EN 511 (021)
- Food contact approved:
- +
- +
- +
Frequently Bought Together