Microscale organic chemistry kit, Quickfit®
Supplier: SciLabware
Certificates
About this item
Microscale shemistry kit that is designed for organic chemistry experiments using milligram quantities of reagents. The components allow the construction of many assemblies including reflux, distillation, moisture sensitive reactions and fractional distillation.
- Comprises 12 items of glassware that all feature 14/10 joint sizes for compatibility- see spare parts list
- Other components compatible with the kit ar listed below under accessories
- All components utilise a combination of standard Quickfit® ground glass joints together with screwthread couplings, sealing 'O' rings and plastic screw caps
- The screwthread connector system eliminates the requirement for grease, saves on space, using clamps and vastly reduces the danger of glassware becoming loose during reactions
Delivery: Supplied in a sturdy Gratnells tray with protective foam insert. Also included in the kit is a bag of spare caps, 'O' rings, sealing discs.
Frequently Bought Together