Order Entry
Denmark
ContactUsLinkComponent
Adenine deaminase
Adenine deaminase
Catalog # 786-428
Supplier:  G-Biosciences
CAS Number:  
undefined
Adenine deaminase
Catalog # 786-428
Supplier:  G-Biosciences
CAS Number:  
Specifications
  • Pk:
    0,25 mL
  • Description:
    Adenine deaminase
  • Environmentally Preferable:
Specifications
About this item
Adenine deaminase converts adenine to hypoxanthine. It is a highly purified recombinant enzyme protein supplied at a 1 µM concentration.