Order Entry
Denmark
ContactUsLinkComponent
Biotin resin
Biotin resin
Catalog # 786-598
Supplier:  G-Biosciences
CAS Number:  
undefined
Biotin resin
Catalog # 786-598
Supplier:  G-Biosciences
CAS Number:  
Specifications
  • Pk:
    5 mL
  • Description:
    Immobilised biotin resin
Specifications
About this item
Immobilised biotin resin is designed for the high affinity chromatography purifications of avidin, streptavidin and neutravidin protein. The resin consists of biotin coupled to 6% cross-linked agarose.
  • Strong affinity for avidin, streptavidin and neutravidin
  • Reusable resin, at least 10 times
  • Covalently coupled to limit leaching