Order Entry
Denmark
ContactUsLinkComponent
Xylene (mixture of isomers) ≥98.5% (by GC) contains ethylbenzene, BAKER ANALYZED® ACS, J.T.Baker®
Xylene (mixture of isomers) ≥98.5% (by GC) contains ethylbenzene, BAKER ANALYZED® ACS, J.T.Baker®
Catalog # 9490-03
Supplier:  Avantor
CAS Number:  
undefined
Xylene (mixture of isomers) ≥98.5% (by GC) contains ethylbenzene, BAKER ANALYZED® ACS, J.T.Baker®
Catalog # 9490-03
Supplier:  Avantor
CAS Number:  

Specifications

  • Pack type:
    Glass bottle, amber, narrow mouth
  • Pk:
    4 L
  • Seq:
    5162
  • Environmentally Preferable:

Specifications

Specification Test Results

  • Meets ACS Reagent Chemical Requirements:
    = 98.5 %
  • For Laboratory,Research,or Manufacturing Use:
    = 98.5 %
  • Assay (m,p,o-xylenes and ethylbenzene) (by GC):
    ≥ 98.5 %
  • Ethylbenzene (C₆H₅C₂H₅):
    ≤ 25 %
  • Color (APHA):
    ≤ 10
  • Residue after Evaporation:
    ≤ 0.0020 %
  • Substances Darkened by H₂SO₄:
    Passes Test
  • Sulfur Compounds (as S):
    ≤ 0.003 %
  • Water (H₂O):
    ≤ 0.02 %

Specification Test Results

About this item

Synonyms: Dimethylbenzene
 :
 :106.17 g/mol
Flash Pt:25
EINECS:215-535-7
UN:1307
 :GHS02
 :GHS07
 :GHS08
Storage Temperature:Ambient
CAS Number:1330-20-7
Formula:C₆H₄(CH₃)₂
MDL Number:MFCD00077264
Boiling Pt:140
Melting Pt:–34 °C
Density:0.86…0.88 g/cm³ (20 °C)
ADR:3 III
Merck Index:13,10136