SAM Methyltransferase assay, SAMfluoro™
Catalog # 786-431
Supplier: G-Biosciences
CAS Number:
Specifications
- Pk:100 Assays
- Description:SAM Methyltransferase Assay Kit, SAMfluoro™
- Tests per kit:100 assays
Specifications
About this item
The SAMfluoro™ assay is a fluorescent, continuous enzyme coupled assay that can continuously monitor SAM-dependent methyltransferases without the use of radioactive labels or endpoint measurements.
- Detection of protein methylation or screening methylation inhibitors
- Continuous enzyme coupled assay for kinetic studies
- Fluorescent, non-radioactive assay
- Supplied with all reagents, including positive control
- Adaptable for enzymes that generate S-adenosylhomocysteine or 5’-methylthioadenosine
Delivery: The kit is supplied with the reagents for 100 assays and AdoHcy as a positive control.