Order Entry
Denmark
ContactUsLinkComponent
Tick tweezers
Tick tweezers
Catalog # OUTITICK
Supplier:  OUTILS RUBIS
undefined
Tick tweezers
Catalog # OUTITICK
Supplier:  OUTILS RUBIS
Specifications
  • Type:
    Angled, very fine tips
  • Description:
    Tweezers, for removing ticks
  • Seq:
    01
  • Pk:
    1
Specifications
About this item
Stainless steel, anti-magnetic. These acid-proof tweezers are ideal for removing ticks. Thanks to the straight, very fine tips that are also angled to the inside, ticks can easily be removed.
  • Also perfect for the removal of thorns and splinters
  • Anti-magnetic and rust-proof surgical steel
  • Made completely by hand - and tested individually under a magnifying glass