Order Entry
Denmark
Orders LinkContactUsLinkComponent
Bin liner holder, Tork W1
Bin liner holder, Tork W1
Bin liner holder, W1, black, for use with the Tork floor stand
Catalog # SCAA206550
Supplier:   Essity
Bin liner holder, Tork W1
Catalog # SCAA206550
Supplier:  Essity
Bin liner holder, Tork W1
Bin liner holder, Tork W1
Bin liner holder, W1, black, for use with the Tork floor stand
Price
442,00 kr
Log in for availability
1 * 1 Items
Product Details & Documents
For use with the Tork Floor Stand (W1), the Tork Bin Liner Holder is a convenient way to store used wipers and cloths when wiping and cleaning whilst moving around the site.
  • Secure bin liner fastening: the bag will not fall off when in use
  • Intutive fitting: easily attached to the floor stand
Specifications
  • Description:
    Bin liner holder, W1, black, for use with the Tork floor stand