Order Entry
Denmark
ContactUsLinkComponent
Alcohol swabs
Alcohol swabs
Catalog # 720-2416
Supplier:  Vitrex Medical A/S
undefined
Alcohol swabs
Catalog # 720-2416
Supplier:  Vitrex Medical A/S

Specifications

  • Description:
    Alcohol swabs, 30×60 mm
  • Pk:
    100

Specifications

About this item

VITREX® alcohol swabs are an easy and convenient way to cleanse the skin and help fight bacteria.

  • Disposable for convenience and safety
  • Contains 70% isopropyl alcohol

Use as an antiseptic skin cleanser before injections and blood sampling.