Order Entry
Denmark
ContactUsLinkComponent
Clamp locks for attaching reaction vessel
Clamp locks for attaching reaction vessel
Catalog # RLAA2124
Supplier:  R L ENTERPRISES
Clamp locks for attaching reaction vessel
Catalog # RLAA2124
Supplier:  R L ENTERPRISES
Specifications
  • For:
    Quickfit® reaction vessels
  • Pk:
    1
Specifications
About this item
Alloy/zinc plated steel construction.
  • Extension rods are secured to the body of the clamp with a socket screw (an allen key is provided)
  • Jaws are heat resistant
Actuated by nickel-plated pummels and twin screw and fitted with stainless steel pivot pins. Each clamp is provided with two extension rods, one for vessels up to 5 L and the other up to 20 L.