Order Entry
Denmark
Orders LinkContactUsLinkComponent
Urine containers, SpecTainer™
Urine containers, SpecTainer™
120 ml, Sterile
Catalog # SIMPC566-120CYS
Supplier:   Simport Scientific
Urine containers, SpecTainer™
Catalog # SIMPC566-120CYS
Supplier:  Simport Scientific
Urine containers, SpecTainer™
Urine containers, SpecTainer™
120 ml, Sterile
Price
kr1,250.00
Log in for availability
1 * 100 Items
Product Details & Documents
PP urine container with tamper evident PE screw cap, ideal for collecting, storing and transporting urine samples.
  • Tamper evident sealing ring for better sample protection
  • Ridges around base offer a better grip during opening and closing
For applications needing the utmost security where sample integrity is of high importance. A great feature of the Spectainer™ I is that it incorporates a unique tamper evident screw cap ensuring peace of mind during transport or storage situations where someone might have tampered with the specimen. For colour coding purposes, place a Capinsert™ on top of closure (see Series T345). Ten different colours are available.
Documents
Specifications
  • Version:
    Sterile
  • Capacity:
    120 ml
  • Seq:
    06