Order Entry
Denmark
Orders LinkContactUsLinkComponent
Meker burners
Supplier:   R L ENTERPRISES
Meker burners
Supplier:  R L ENTERPRISES
Certificates
9 Options Available Below
Product Details & Documents
Nickel plated brass. The top of the burner is fitted with a perforated steel head, which gives multiple small but very hot flames. It is not fitted with an air regulator. Burners have diecast base painted in a blue chemically resistant acrylic finish.
Product Family Options
Product Information
  • For
    Head Ø
    Availability
    Price
  • RLAA4101/21
    Meker burner, with head Ø 20 mm
    Meker burner, with head Ø 20 mm
    LPG
    20 mm
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 922,00 kr
    922,00 kr
    1 * 1 Items
       922,00 kr
  • RLAA4101/31
    Meker burner, with head Ø 24 mm
    Meker burner, with head Ø 24 mm
    LPG
    24 mm
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 1.070,00 kr
    1.070,00 kr
    1 * 1 Items
       1.070,00 kr
  • RLAA4101/32
    Meker burner, with head Ø 30 mm
    Meker burner, with head Ø 30 mm
    Natural gas
    30 mm
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 1.380,00 kr
    1.380,00 kr
    1 * 1 Items
       1.380,00 kr
  • RLAA4101/3
    Meker burner, with head Ø 24 mm
    Meker burner, with head Ø 24 mm
    Coal
    24 mm
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 1.070,00 kr
    1.070,00 kr
    1 * 1 Items
       1.070,00 kr
  • RLAA4101/41
    Meker burner, with head Ø 30 mm
    Meker burner, with head Ø 30 mm
    LPG
    30 mm
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 1.380,00 kr
    1.380,00 kr
    1 * 1 Items
       1.380,00 kr
  • RLAA4101/4
    Meker burner, with head Ø 30 mm
    Meker burner, with head Ø 30 mm
    Coal
    30 mm
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 1.380,00 kr
    1.380,00 kr
    1 * 1 Items
       1.380,00 kr
  • RLAA4101/51
    Meker burner, with head Ø 43 mm
    Meker burner, with head Ø 43 mm
    LPG
    43 mm
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 3.650,00 kr
    3.650,00 kr
    1 * 1 Items
       3.650,00 kr
  • RLAA4101/52
    Meker burner, with head Ø 43 mm
    Meker burner, with head Ø 43 mm
    Natural gas
    43 mm
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 3.650,00 kr
    3.650,00 kr
    1 * 1 Items
       3.650,00 kr
  • RLAA4101/5
    Meker burner, with head Ø 43 mm
    Meker burner, with head Ø 43 mm
    Coal
    43 mm
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 3.650,00 kr
    3.650,00 kr
    1 * 1 Items
       3.650,00 kr
Anbefalinger