Order Entry
Denmark
ContactUsLinkComponent
Surface Cleaner, aerosol spray, Super-Degreaser
Surface Cleaner, aerosol spray, Super-Degreaser
Catalog # KING050502
Supplier:  KING DISPOSABLES
undefined
Surface Cleaner, aerosol spray, Super-Degreaser
Catalog # KING050502
Supplier:  KING DISPOSABLES
Specifications
  • Description:
    Super-Degreaser, aerosol spray surface cleaner,
  • Packed:
    400 ml aerosol can
  • Pk:
    1
Specifications
About this item
Kitchen degreaser for removing fats and burnt on fat residues from ceramics, microwave interiors, glass oven doors, even barbecues.
  • Does not contain caustic soda
  • Compatible with metals, ceramics plastics and glass