Glove liners, PU Fingertip, A121
Supplier: Portwest
Certificates
About this item
These knitted glove liners made of nylon are suitable for use in microelectronics, precision assembly and inspection.
- Fingertips with dipped PU coating for protection against nicks and cuts
- High abrasion resistance
Gauge: 13
Colour: White
Cat. I
Certifications: Cat. I
Specifications
- Catalog No:
- PRTWA121WHRS
- PRTWA121WHRXS
- PRTWA121WHRXL
- PRTWA121WHRM
- PRTWA121WHRL
- Model:
- A121
- A121
- A121
- A121
- A121
- Ambidextrous / hand-specific:
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Disposable / reusable:
- Reusable
- Reusable
- Reusable
- Reusable
- Reusable
- Gauge:
- 13
- 13
- 13
- 13
- 13
- Cuff style:
- Knit wrist
- Knit wrist
- Knit wrist
- Knit wrist
- Knit wrist