Adhesive PCR foil seals, QuickSeal Foil PCR ULTRA™
Supplier: iST Scientific Ltd
Certificates
About this item
Pierceable, peelable barrier foil for PCR and sample storage.
- Seal integrity range: −70 to +120 °C
- Available sterile or non sterile