Immersion thermostats
About this item
Immersion thermostats form the basis of many different combinations with polycarbonate or stainless steel bath vessels. All models are fitted with a powerful pressure/suction pump and meet the requirements of Safety Class III (FL) for use with flammable fluids. Temperatures down to −30 °C can be achieved in combination with a refrigerated bath. A screw clamp is included allowing the thermostats to be easily mounted on any vessels. A pump adapter for external temperature control and cooling coils for a cooling water connection are available as optional accessories.
- Pilot ONE® is interchangeable and can be used as a remote control (with an optional data cable)
- The controller can be upgraded with enhanced functionality at any time (Exclusive and Professional E-grade® packages), basic version features 11 languages (German, English, French, Italian, Spanish, Portuguese, Czech, Polish, Russian, Chinese and Japanese)
- The temperature resolution can be displayed to 0,1 or 0,01 °C, display can be customised to the user's preferences and important information e.g. set point, actual internal or process temperatures can be displayed in a larger format
- Powerful variable speed pressure/suction pump with variable pressure control (VPC)
- User can set visual and acoustic alarms limiting the temperature range, and program the unit with its clock and calendar functions for automatic startup
Maximum pump capacities:
Pilot ONE® (CC-E): 27 L/min, 0,7 bar (pressure), 22 L/min, 0,4 bar suction
Pilot ONE® (CC-E xd): 22 L/min, 0,4 bar (pressure), 17 L/min, 0,25 bar suction
Modern and easy to use microprocessor controller with a large temperature display limited to essential functions only. MPC controller features over-temperature alarm and low liquid level protection. All MPC models also feature a powerful and suction pump for optimum performance.
Maximum pump capacities:
Pilot ONE® (CC-E): 27 l/min, 0,7 bar, 0,4 bar suction
Pilot ONE® (CC-E xd): 22 l/min, 0,4 bar, 0,4 bar suction
MPC-E: 20 l/min, 0,2 bar, 0,18 bar suction
Certifications: Safety Class III (FL) (DIN 12876) for use with flammable fluids
Ordering information: Accessories included. A pump adapter for external temperature control and cooling coils for a cooling water connection are available as optional accessories.
Caution: *Note: Working temperature range can be lowered to: −30 to +200 °C if an external cooling device is used.
2 Options Available Below
Product Details & Documents
Immersion thermostats form the basis of many different combinations with polycarbonate or stainless steel bath vessels. All models are fitted with a powerful pressure/suction pump and meet the requirements of Safety Class III (FL) for use with flammable fluids. Temperatures down to −30 °C can be achieved in combination with a refrigerated bath. A screw clamp is included allowing the thermostats to be easily mounted on any vessels. A pump adapter for external temperature control and cooling coils for a cooling water connection are available as optional accessories.
- Pilot ONE® is interchangeable and can be used as a remote control (with an optional data cable)
- The controller can be upgraded with enhanced functionality at any time (Exclusive and Professional E-grade® packages), basic version features 11 languages (German, English, French, Italian, Spanish, Portuguese, Czech, Polish, Russian, Chinese and Japanese)
- The temperature resolution can be displayed to 0,1 or 0,01 °C, display can be customised to the user's preferences and important information e.g. set point, actual internal or process temperatures can be displayed in a larger format
- Powerful variable speed pressure/suction pump with variable pressure control (VPC)
- User can set visual and acoustic alarms limiting the temperature range, and program the unit with its clock and calendar functions for automatic startup
Maximum pump capacities:
Pilot ONE® (CC-E): 27 L/min, 0,7 bar (pressure), 22 L/min, 0,4 bar suction
Pilot ONE® (CC-E xd): 22 L/min, 0,4 bar (pressure), 17 L/min, 0,25 bar suction
Modern and easy to use microprocessor controller with a large temperature display limited to essential functions only. MPC controller features over-temperature alarm and low liquid level protection. All MPC models also feature a powerful and suction pump for optimum performance.
Maximum pump capacities:
Pilot ONE® (CC-E): 27 l/min, 0,7 bar, 0,4 bar suction
Pilot ONE® (CC-E xd): 22 l/min, 0,4 bar, 0,4 bar suction
MPC-E: 20 l/min, 0,2 bar, 0,18 bar suction
Certifications: Safety Class III (FL) (DIN 12876) for use with flammable fluids
Ordering information: Accessories included. A pump adapter for external temperature control and cooling coils for a cooling water connection are available as optional accessories.
Caution: *Note: Working temperature range can be lowered to: −30 to +200 °C if an external cooling device is used.
Product Family Options
Product Information
- TypeTemperature RangeTemp. stabilityHeating capacityW×D×HWeightAvailabilityPrice
- CC-E(−30) 25...200 °C±0,01 °C1,5 - 2,1 kW132×159×315 mm4,0 kgLog in for availability1 * 1 Items15.420,00 kr
- CC-E xd(−30) 25...200 °C±0,01 °C1,5 - 2,1 kW132×159×360 mm5,0 kgLog in for availability1 * 1 Items18.690,00 kr
Optional upgrade
Product Information
- Catalog #DescriptionAvailabilityPrice
Water baths, transparent polycarbonate
Product Information
- Catalog #Bath W×D×HTypeCapacityW×D×HTemperature RangeAvailabilityPrice
- 480×180×312 mmPolycarbonate bath, 130A-E30 L500×200×322 mm≤100 °CLog in for availability1 * 1 Items7.040,00 kr
Water baths, insulated stainless steel
Product Information
- Catalog #Bath W×D×HTypeCapacityW×D×HTemperature RangeAvailabilityPrice
- 290×320×200 mmStainless steel bath, 215B15 L350×375×256 mm≤200 °CLog in for availability1 * 1 Items6.750,00 kr
Water baths, insulated stainless steel Variostat®
Product Information
- Catalog #Bath W×D×HTypeCapacityW×D×HTemperature RangeAvailabilityPrice
- 468×320×165 mmVariostat® bath, stainless steel22 L520×370×205 mm≤200 °CLog in for availability1 * 1 Items8.800,00 kr