Order Entry
Denmark
Orders LinkContactUsLinkComponent
Extraction hood, SICCO
 :   Bohlender
Extraction hood, SICCO
 :  Bohlender
Extraction hood, SICCO
Extraction hood, SICCO
Anodized aluminium frame with panels made of polycarbonate.
  • Transparent and shatter-proof panels
  • Robust and light-weight aluminium construction
  • Standard connection DN 160 for aspirator tube
  • Temperature resistance: from −20 to +70 °C
Top panel with integrated connector for exhaust system offers a good chemical resistance. These extractor hoods are used for a diverse range of industrial applications such as handling of varnish and combustible liquids.
Product Family Options
  • Description
    W×D×H ext.
    W×D×H int.
    Weight
  • Extractor hood, opening dimension 545×600 mm
    900×600×900 mm
    860×560×880 mm
    20
    1 * 1 Items
    1 * 1 Items - 20.320,00 kr
    20.320,00 kr
    1 * 1 Items
       20.320,00 kr
  • Extractor hood, opening dimension 850×600 mm
    1200×600×900 mm
    1160×560×880 mm
    23
    1 * 1 Items
    1 * 1 Items - 25.080,00 kr
    25.080,00 kr
    1 * 1 Items
       25.080,00 kr