Order Entry
Denmark
Orders LinkContactUsLinkComponent
Disposable powder samplers, SteriWare®
Disposable powder samplers, SteriWare®
Disposable powder samplers, SteriWare®
Supplier:  Sampling Systems
Certificates
Disposable powder samplers, SteriWare®
Disposable powder samplers, SteriWare®
2 Options Available Below
Product Details & Documents
Designed to collect single large volume sample from a single known point of free flowing powders from drums, kegs, and most containers.
  • Single use with gamma irradiation
  • Assembled and packed in a cleanroom environment - Class 100000
  • BSE/TSE free as well as full batch traceability
 : FDA and EU 10/2011 conformity, ATEX rated.
Product Family Options
Product Information
  • Material
    Capacity
    Ø
    Length
    Sterile
    Availability
    Price
  • Disposable samplers
    HDPE
    100 ml
    40 mm
    1000 mm
    +
    Log in for availability
    Log in for availability
    1 * 20 Items
    1 * 20 Items - 5.030,00 kr
    5.030,00 kr
    1 * 20 Items
       5.030,00 kr
  • SAMP8240H-1000
    Disposable samplers
    Disposable samplers
    HDPE
    100 ml
    40 mm
    1000 mm
    Log in for availability
    Log in for availability
    1 * 20 Items
    1 * 20 Items - 3.840,00 kr
    3.840,00 kr
    1 * 20 Items
       3.840,00 kr