Hand disinfectants, Septigel+
Catalog # 115-0627
Leverandør: LABORATOIRES PRODENE KLINT
Specifikationer
- Description:Hand disinfectant, Septigel+, bottle
- Pk:24
Specifikationer
Om denne vare
Septigel+ is a skin-friendly, moisturising and fast acting hand disinfectant for everyday use.
- Bactericidal: Active on Salmonella enterica, Listeria monocytogenes
- Virucidal: Active on HIV, Rotavirus, Herpes virus, BVDV (Hepatitis C indicator), PRV (Hepatitis B indicator), MNV (murine norovirus, model virus for the study of human noroviruses), Poliovirus, Adenovirus
- Fungicidal: Candida albicans, Aspergillus niger
- Tuberculocidal: Mycobacterium terrae
- Contains ethanol, isopropanol, glycerine
- No rinsing required
- Colourant- and perfume-free
- Hypoallergenic