Order Entry
Denmark
ContactUsLinkComponent
Formic acid ≥99%, HiPerSolv CHROMANORM® for LC-MS
Flammable
Corrosive
Toxic
Formic acid ≥99%, HiPerSolv CHROMANORM® for LC-MS
Supplier:  VWR Chemicals
Certificates
undefined
Flammable
Corrosive
Toxic
Formic acid ≥99%, HiPerSolv CHROMANORM® for LC-MS
Supplier:  VWR Chemicals
Certificates

Specification Test Results

  • Assay (on anhydrous substance):
    Min. 99 %
  • Evaporation residue:
    Max. 0.0001 %
  • Ca (Calcium) (0.1 %):
    Max. 0.2 ppm
  • Fe (Iron) (0.1%):
    Max. 0.2 ppm
  • K (Potassium) (0.1 %):
    Max. 0.2 ppm
  • Mg (Magnesium) (0.1 %):
    Max. 0.2 ppm
  • Na (Sodium) (0.1 %):
    Max. 0.5 ppm
  • Transmittance (225 nm) (0.1 %):
    Min. 20 %
  • Transmittance (235 nm) (0.1 %):
    Min. 50 %
  • Transmittance (240 nm) (0.1 %):
    Min. 80 %
  • Transmittance (250 nm) (0.1 %):
    Min. 95 %
  • Transmittance (260 nm) (0.1 %):
    Min. 99 %
  • Suitability for LC-MS (0.1 %):
    Passes test

Specification Test Results

Frequently Bought Together