Disinfectants, ClearKlens DE
: Diversey
ClearKlens DE is a ready to use sterile disinfectant with 70% ethanol and 30% WFI (water for injection). It has the broad spectrum of activity against both gram-positive and gram-negative bacteria, yeasts and fungi. Can be used for the disinfection of work surfaces and process equipment in cleanrooms. Designed for spraying and wiping applications.
- 0,2 μm filtered, gamma irradiated (900 ml) or sterile filled (5 L pack size) and double-bagged
- Detex-labelled for proof of irradiation (900 ml)
- Supplied with Certificates of Analysis, irradiation, endotoxin and sterility where applicable
- 900 ml trigger spray pack incorporates anti-suck technology
: Bactericide EN 1276; Fungicide EN 1650; Bactericide and Fungicide EN 13697.
: Use biocides safely. Always read the label and product information before use.
Frequently Bought Together