SPE columns, Supra-Poly® XWP
Supplier: PerkinElmer
Certificates
About this item
SPE Supra-Poly® extreme capacity wide pore (XWP) are polymer based columns, polystyrene-divinylbenzene (PSDVB). Ideal for extracting polar and non-polar compounds when analysing drugs and drug metabolites in biological fluids.
Frequently Bought Together