Order Entry
Denmark
Orders LinkContactUsLinkComponent
Disposable cuvettes
Disposable cuvettes
Supplier:   SEMADENI
Disposable cuvettes
Supplier:  SEMADENI
Certificates
Disposable cuvettes
3 Options Available Below
Product Details & Documents
Disposable cuvettes for photometric measurements in the visible range (350 to 800 nm).
Product Family Options
Product Information
  • Type
    Range
    Height
    Volume
    Path length
    Window material
    Availability
    Price
  • Semi-micro
    350 – 800 nm
    45 mm
    1,5 ml
    10 mm
    PS
    Log in for availability
    Log in for availability
    1 * 100 Items
    1 * 100 Items - kr399.00
    kr399.00
    1 * 100 Items
       kr399.00
  • Macro
    350 – 800 nm
    45 mm
    4,5 ml
    10 mm
    PS
    Log in for availability
    Log in for availability
    1 * 100 Items
    1 * 100 Items - kr310.00
    kr310.00
    1 * 100 Items
       kr310.00
  • Macro
    350 – 800 nm
    45 mm
    4,5 ml
    10 mm
    PMMA
    Log in for availability
    Log in for availability
    1 * 100 Items
    1 * 100 Items - kr566.00
    kr566.00
    1 * 100 Items
       kr566.00
Accessories
Product Information
  • Catalog #
    Availability
    Price
  • LDPE caps for cuvettes with square opening
    LDPE caps for cuvettes with square opening
    SEMA2533
    Log in for availability
    1 * 100 Items - 139,00 kr