Order Entry
Denmark
Orders LinkContactUsLinkComponent
Safety spectacles, Rush+
Safety spectacles, Rush+
Supplier:   Bollé Safety
Safety spectacles, Rush+
Supplier:  Bollé Safety
Certificates
Safety spectacles, Rush+
Safety spectacles, Rush+
10 Options Available Below
Product Details & Documents
The RUSH+ model adopts an ultra-sporty design with its co-injected and ultra-flexible temples and is 100% customisable.
  • Upper protection
  • Co-injected, ultra-flexible temples
  • Non-slip adjustable bridge
Available with Twilight technology: ESP with a double anti-fog coating (on both sides of the lenses). With Platinum technology a permanent coating on both sides of the lens gives them a high resistance to scratching (1,4 cd/m²), withstands most aggressive chemicals, and delays the onset of fogging for 2 minutes. Designed for use in low light conditions, it improves contrast. The light transmission rate is ideal for use outdoors, particularly early morning and late evening. B-Twilight filters 76% of blue light.
Certifications: EN 166 FT
Product Family Options
Product Information
  • Type
    Colour
    Lenses
    Availability
    Price
  • Anti-fog, anti-scratch
    Grey/grey
    CSP PC
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 112,00 kr
    112,00 kr
    1 * 1 Items
       112,00 kr
  • Anti-fog, anti-scratch
    Red/black
    Clear PC
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 190,00 kr
    190,00 kr
    1 * 1 Items
       190,00 kr
  • Anti-UV (5-3,1), anti-fog, anti-scratch
    Red/black
    Smoke PC
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 113,00 kr
    113,00 kr
    1 * 1 Items
       113,00 kr
  • Anti-UV (2C-1,2), anti-fog, anti-scratch
    Blue/black
    Clear PC
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 102,00 kr
    102,00 kr
    1 * 1 Items
       102,00 kr
  • Anti-UV (2C-1,2), anti-fog, anti-scratch
    Green/black
    Clear PC
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 102,00 kr
    102,00 kr
    1 * 1 Items
       102,00 kr
  • Anti-UV (2C-1,2), anti-fog, anti-scratch
    Orange/black
    Clear PC
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 102,00 kr
    102,00 kr
    1 * 1 Items
       102,00 kr
  • Anti-UV (2C-1,2), anti-fog, anti-scratch
    Red/black
    Clear PC
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 102,00 kr
    102,00 kr
    1 * 1 Items
       102,00 kr
  • Anti-fog, anti-scratch
    Blue/grey
    Smoke PC
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 113,00 kr
    113,00 kr
    1 * 1 Items
       113,00 kr
  • Anti-fog, anti-scratch
    Clear/red
    Clear PC
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 102,00 kr
    102,00 kr
    1 * 1 Items
       102,00 kr
  • Twilight, anti-UV (5-2), anti-fog, anti-scratch
    Grey/black
    Brown PC
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 113,00 kr
    113,00 kr
    1 * 1 Items
       113,00 kr