Cleanroom wipes, sterile, Klerwipe® Sterile
These low particulate wipes are made from a binder free, hydroentangled cellulose/polyester blend. Suitable for use in controlled environments such as isolators and laminar flow cabinets.
- Highly absorbent: Ideal for applying cleaning solutions and disinfectants and for cleaning up spills
- Do not contain any chemical binders and do not leave undesirable residues
- Packed in small quantities to avoid the need to compromise the sterility of a pack, once opened
The dry wipes are ideal for all cleaning and disinfection procedures. The amount of cleaning agent can be varied to suit the particular purpose.
Frequently Bought Together