Order Entry
Denmark
ContactUsLinkComponent
Desiccator cabinet, SICCO Super Big Star
Desiccator cabinet, SICCO Super Big Star
Catalog # BOHLV1935-07
Supplier:  Bohlender
undefined
Desiccator cabinet, SICCO Super Big Star
Catalog # BOHLV1935-07
Supplier:  Bohlender

Specifications

  • Weight:
    18 kg
  • W×D×H ext.:
    560×580×560 mm
  • Description:
    Super Big Star (140 l)
  • Pk:
    1
  • Shelves:
    2
  • W×D×H int.:
    495×540×500 mm

Specifications

About this item

SICCO desiccators are designed for storing and drying humidity sensitive products using silica gel. The tight fitting door protects the contents from contamination in the atmosphere.

  • Reinforced aluminium frame with acrylic panels
  • Door with magnetic catch and circular rubber seal
  • Non slip rubber feet
  • Easy to read electronic hygrometer indicates minimum and maximum temperature and humidity values
  • Variable height shelves made of stainless steel
  • Telescopic shelves
  • Temperature resistance: –20 to +70 °C

The controlled environment inside the desiccator makes it ideal for storing reference materials, retained samples, metallographic specimens and DNA samples.

Delivery: Supplied with two stainless steel shelves, tray and hygrometer and hose (5 m) with two quick couplings for gas filling, usable with up to eight shelves.