Order Entry
Denmark
Orders LinkContactUsLinkComponent
Saccharometers
Saccharometers
Supplier:   Amarell
Saccharometers
Supplier:  Amarell
Certificates
Saccharometers
6 Options Available Below
Product Details & Documents
Saccharometers with reference temperature of 20 °C.
Product Family Options
Product Information
  • Range
    Division
    Length
    Availability
    Price
  • 0 - 10
    0,1
    260 mm
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 272,00 kr
    272,00 kr
    1 * 1 Items
       272,00 kr
  • 10 - 20
    0,1
    260 mm
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 272,00 kr
    272,00 kr
    1 * 1 Items
       272,00 kr
  • 20 - 30
    0,1
    260 mm
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 272,00 kr
    272,00 kr
    1 * 1 Items
       272,00 kr
  • 30 - 40
    0,1
    260 mm
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 272,00 kr
    272,00 kr
    1 * 1 Items
       272,00 kr
  • 40 - 50
    0,1
    260 mm
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 272,00 kr
    272,00 kr
    1 * 1 Items
       272,00 kr
  • 60 - 80
    1,0
    200 mm
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 288,00 kr
    288,00 kr
    1 * 1 Items
       288,00 kr