SMD tweezers, Erem®
: Basan
Stainless steel. These tweezers are ideal for the gripping and holding of cylindrical components. Available in straight and angled versions.
- Non rusting, heat resistant, and anti-acid proof surface
- Anti-magnetic
- With blunted edges to prevent damage to printed-circuit boards
Frequently Bought Together