Order Entry
Denmark
Orders LinkContactUsLinkComponent
Capillary columns for specific EPA methods, Trace™
Capillary columns for specific EPA methods, Trace™
Capillary columns for specific EPA methods, Trace™
Capillary columns for specific EPA methods, Trace™
These columns are designed to perform specific EPA methodologies. These low-bleed and temperature-stable columns have been designed, manufactured and tested to provide consistent, reliable results from run-to-run and batch-to-batch.
  • Low bleed, even at elevated temperatures
  • Outstanding robustness for difficult separations
  • Reliable and reproducible performance
Applications: Volatile organic compounds (VOCs), pesticides, brominated flame retardants, explosives.
Product Family Options
  • Column Format
    Phase
    Film thickness
    Ø int.
    Length
  • Capillary
    Trace™ TR-525
    0,25 µm
    0,25 mm
    30 m
    1 * 1 Items
    1 * 1 Items - 8.260,00 kr
    8.260,00 kr
    1 * 1 Items
       8.260,00 kr
  • Capillary
    Trace™ TR-8270
    0,50 µm
    0,25 mm
    30 m
    1 * 1 Items
    1 * 1 Items - 8.470,00 kr
    8.470,00 kr
    1 * 1 Items
       8.470,00 kr
  • Capillary
    Trace™ TR-524
    1,00 µm
    0,18 mm
    20 m
    1 * 1 Items
    1 * 1 Items - 7.240,00 kr
    7.240,00 kr
    1 * 1 Items
       7.240,00 kr