Transport container, round
Supplier: SARSTEDT
Certificates
About this item
These mailing containers are made of PP. Suitable for the transport of diagnostic samples.
- Leakproof at a pressure difference of 95 kP
- Without cap
Certifications: Compliance with the P650/ADR packaging instructions.
Frequently Bought Together