Order Entry
Denmark
ContactUsLinkComponent
Protective barrier creams, Sineprint®
Protective barrier creams, Sineprint®
Catalog # 115-1755
Supplier:  FAWECO HAUTSCHUTZ
undefined
Protective barrier creams, Sineprint®
Catalog # 115-1755
Supplier:  FAWECO HAUTSCHUTZ
Specifications
  • Description:
    Protective barrier cream, Sineprint®, 100 ml Tube
  • Pk:
    12
Specifications
About this item
White, non-greasy barrier cream (oil/water emulsion) for use in workplaces where greasy residue must be avoided, such as in the production of electronics, final inspections, paint shops, or in the paper industry.
  • No greasy residue and absorbs quickly
  • Silicone-free, scented
  • Neutral pH value, contains no mineral oil
Caution: *The product(s) on this page are outside the scope of the EU Directive 2019/904