Photographic trays
Supplier: DELTALAB
Certificates
About this item
Photographic trays made of PVC.
- Temperature resistance from −20 to +80 °C
With ribbed interior surface.
Frequently Bought Together
Photographic trays made of PVC.
With ribbed interior surface.