Order Entry
Denmark
ContactUsLinkComponent
Small molecular model: DNA/RNA
Small molecular model: DNA/RNA
Catalog # BINH1005302
Supplier:  3B Scientific
undefined
Small molecular model: DNA/RNA
Catalog # BINH1005302
Supplier:  3B Scientific

Specifications

  • Weight:
    8 g
  • Description:
    Small molecular model of DNA/RNA
  • W×D×H:
    200×300×5 mm
  • Pk:
    1

Specifications

About this item

The small DNA/RNA model shows the double helix and how the molecules split at the centre of the base pairs. It includes:

  • 30 coloured tubes – Guanine red, cytosine green, thymine blue, adenine gray and uracil white
  • 12 white two prong centre representing the hydrogen bond between the base pairs
  • 28 black trigonal atom centres showing sugar
  • 25 red two prong atom centres, which represent phosphate groups
  • 50 yellow tubes that link the phosphate groups to the sugar rings
Ordering information: Includes a wooden base with support rod.