Order Entry
Denmark
ContactUsLinkComponent
Six mounted vertebrae
Six mounted vertebrae
Catalog # BINH1000147
Supplier:  3B Scientific
undefined
Six mounted vertebrae
Catalog # BINH1000147
Supplier:  3B Scientific

Specifications

  • Description:
    Six mounted vertebrae
  • Pk:
    1

Specifications

About this item

Consisting of atlas, axis, another cervical vertebra, two thoracic vertebrae with inter-vertebral discs and one lumbar vertebra.

  • On stand
  • Removable

Height: 22 cm

Weight: 0,3 kg