Order Entry
Denmark
ContactUsLinkComponent
Beaker cups, PP, medicine
Beaker cups, PP, medicine
Catalog # 216-4220
Supplier:  DELTALAB
Beaker cups, PP, medicine
Catalog # 216-4220
Supplier:  DELTALAB

Specifications

  • Ext.Ø:
    35 mm
  • Capacity:
    25 ml
  • Height:
    40 mm
  • Pk:
    100

Specifications

About this item

PE, translucent.

  • Good chemical resistance
  • Good temperature resistance

Robust graduated medicine measures.