BOD apparatus, BODTrak™II
BODTrak™II, respirometric BOD apparatus, EU-plug
Catalog # HACH2952400
Supplier: Hach
Jump to:
Product Details & Documents
The BODTrak™ II apparatus is easy to set up and allows for quick sample preparation for BOD analysis.
- Standalone operation
- Large graphic display that continuously updates results that can be displayed as a curve
- Magnetic stirring of each bottle for more rapid results than dilution method
- Selectable range and test duration
Delivery information: Supplied with six bottles, six magnetic stir bars, six seal cups, spatula scoop, 50 BOD nutrient buffer pillows and potassium hydroxide pellets
Specifications
- Description:BODTrak™II, respirometric BOD apparatus, EU-plug
- Interfaces:RS232
- W×D×H of system:289×260×98 mm
- Cat. No.:HACH2952400
- Capacity:6×492 ml bottles
- Model:BODTrak™II
- Range:Selectable: 0 to 35, 0 to 70, 0 to 350, or 0 to 700 mg/l BOD
- Weight:4 kg
- Resolution:1 mg/l
Accessories
Product Information
- Catalog #AvailabilityPrice
- Respirometric BOD nutrient buffer solution pillowsRespirometric BOD nutrient buffer solution pillowsHACH2962266Log in for availability1 * 50 Items - 376,00 kr
Product Information
- Catalog #AvailabilityPrice
- Standard methods, for water and waste water analysisStandard methods, for water and waste water analysisHACH2270800Log in for availability1 * 1 Items - 6.120,00 kr
Product Information
- Catalog #AvailabilityPrice
- Storage bottle, glass, amber, BODTrak™II
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



