Spoons, double-ended
Supplier: HAMMACHER KARL
Jump to:
About this item
Double-ended spoons made from WIRONIT steel (18/12 stainless steel with 18% chrome and 10 to 12% nickel).
3 Options Available Below
Product Details & Documents
Double-ended spoons made from WIRONIT steel (18/12 stainless steel with 18% chrome and 10 to 12% nickel).
Product Family Options
Product Information