UV pocket flashlight
LED UV pocket flashlight, beam distance 50 m, Industries and laboratory, 395/410 nm
Catalog # SIMELD7290
Supplier: Industrial Physics
Jump to:
Product Details & Documents
This UV pocket flashlight is designed to detect contaminations that react under UV illumination and can not be seen with naked eye such as some organic fats and alkaline contaminants. They are also suitable to inspect the cleanliness of steel prior to painting.
W×D: 32×126 mm
Delivery information: Supplied with 3× AAA alkaline batteries.
Documents
TQC Company Video
Videos
Specifications
- For:Industries and laboratory
- Description:LED UV pocket flashlight, beam distance 50 m
- Wavelength:395/410 nm
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



