Single channel pipettes, variable volume, MLA Digital Adjustable
Digital adjustable pipettes are an ideal choice for laboratories that perform testing requiring multiple or non standard volumes. They are made of engineered aluminum alloy and stainless steel for maximum durability and performance.
- Three models to cover volume range of 10 to 1000 μl
- Lower spring forces to minimise thumb muscle activity
- Simple dial-in settings with 'click stops' provide fast and easy volume selection
- Streamlined shape and smooth, singlestroke operation make these pipettes easy to use
All pipettes are engraved with serial numbers to facilitate improved traceability and calibration recordkeeping.
Digital adjustable pipettes are an ideal choice for laboratories that perform testing requiring multiple or non standard volumes. They are made of engineered aluminum alloy and stainless steel for maximum durability and performance.
- Three models to cover volume range of 10 to 1000 μl
- Lower spring forces to minimise thumb muscle activity
- Simple dial-in settings with 'click stops' provide fast and easy volume selection
- Streamlined shape and smooth, singlestroke operation make these pipettes easy to use
All pipettes are engraved with serial numbers to facilitate improved traceability and calibration recordkeeping.
Product Family Options
- VolumeAccuracyImprecisionColour code
- 10 - 50 µl±5 - ±2 %<2,6 - <0,80 %Green1 * 1 Items4.890,00 kr
- 50 - 200 µl±2 - ±1 %<0,80 - <0,50 %Red1 * 1 Items4.890,00 kr
- 200 - 1000 µl±1 %<0,50 - <0,60 %Grey1 * 1 Items4.890,00 kr
Accessories
- Description
- Description
- Description
- Description
- Description
- Description