ABTS (Diammonium 2,2'-azinobis[3-ethyl-2,3-dihydrobenzothiazole-6-sulphonate]) ≥98.0% (by HPLC, titration analysis) spectrophotometric reagent for free chlorine
Catalog # TCIAA5200-1G
Supplier: TCI
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:1 g
- Seq:3047
Specifications
Specification Test Results
- Appearance:White to Light yellow to Green powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (Neutralization titration):min. 98.0 %
- suitability for Chlorin-analysis:Abs min. 0.11 (414 to 420nm) in the presence of Chloride ion (0.1 ppm)
Specification Test Results
About this item
:
:548.68 g/mol
EINECS:250-396-6
:GHS07
Storage Temperature:Refrigerator
CAS Number:30931-67-0
Formula:C₁₈H₂₄N₆O₆S₄
MDL Number:MFCD00010404