Order Entry
Denmark
Orders LinkContactUsLinkComponent
Amphotericin B

Amphotericin B

Irritant
Supplier:   Cayman Chemical
Amphotericin B
Supplier:  Cayman Chemical
Certificates

3 Options Available Below

Product Details & Documents
Synonyms:  (1R,3S,5R,6R,9R,11R,15S,16R,17R,18S,19E,21E,23E,25E,27E,29E,31E,33R,35S,36R,37S)-33-(((2R,3S,4S,5S,6R)-4-Amino-3,5-dihydroxy-6-methyltetrahydro-2H-pyran-2-yl)oxy)-1,3,5,6,9,11,17,37-octahydroxy-15,16,18-trimethyl-13-oxo-14,39-dioxabicyclo[33.3.1]nonatriaconta-19,21,23,25,27,29,31-heptaene-36-carboxylic acid
  • CAS Number:1397-89-3
  • Formula:C₄₇H₇₃NO₁₇
  • MDL Number:MFCD00877763
  • Molecular Weight:924.08 g/mol
  • Storage Temperature:Freezer
  • EINECS:215-742-2
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Glass bottle for solids
    500 mg
    Log in for availability
    Log in for availability
    1 * 500 mg
    1 * 500 mg - 550,00 kr
    550,00 kr
    1 * 500 mg
       550,00 kr

  • Glass bottle for solids
    1 g
    Log in for availability
    Log in for availability
    1 * 1 g
    1 * 1 g - 824,00 kr
    824,00 kr
    1 * 1 g
       824,00 kr

  • Glass bottle for solids
    5 g
    Log in for availability
    Log in for availability
    1 * 5 g
    1 * 5 g - 2.720,00 kr
    2.720,00 kr
    1 * 5 g
       2.720,00 kr