

4-Methylstyrene ≥96.0% (by GC) stabilised
Supplier: TCI
Certificates
About this item
CAS Number:622-97-9
:
Formula:C₉H₁₀
MDL Number:MFCD00008621
:118.18 g/mol
Boiling Pt:175°C
Melting Pt:–34 °C
Density:0,9383 g/cm³ (0 °C)
Flash Pt:45°C
UN:2618
ADR:3,III
:Stabilised with 4-tert-butylpyrocatechol (TBC)
EINECS:210-762-8
:GHS02
:GHS07
Storage Temperature:Refrigerator
Specification Test Results
- Appearance:Colorless to Almost colorless clear liquid
- Purity (GC):min. 96.0 %
Specification Test Results
Frequently Bought Together