Order Entry
Denmark
Orders LinkContactUsLinkComponent
Spermidine

Spermidine

Supplier:   Cayman Chemical
Spermidine
Supplier:  Cayman Chemical
Certificates

4 Options Available Below

Product Details & Documents
Synonyms:  N1-(3-Aminopropyl)butane-1,4-diamine, N-(3-Aminopropyl)-1,4-diaminobutane, 4-Azaoctamethylenediamine
  • CAS Number:124-20-9
  • Formula:C₇H₁₉N₃
  • MDL Number:MFCD00008229
  • Molecular Weight:145.25 g/mol
  • Merck Index:12,08893
  • Boiling Pt:246 °C (1013 hPa)
  • Melting Pt:20…25 °C
  • Density:0.925 g/cm³ (20 °C)
  • EINECS:204-689-0
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Glass bottle for solids
    1 g
    Log in for availability
    Log in for availability
    1 * 1 g
    1 * 1 g - 395,00 kr
    395,00 kr
    1 * 1 g
       395,00 kr

  • Glass bottle for solids
    5 g
    Log in for availability
    Log in for availability
    1 * 5 g
    1 * 5 g - 1.050,00 kr
    1.050,00 kr
    1 * 5 g
       1.050,00 kr

  • Glass bottle for solids
    10 g
    Log in for availability
    Log in for availability
    1 * 10 g
    1 * 10 g - 1.860,00 kr
    1.860,00 kr
    1 * 10 g
       1.860,00 kr

  • Glass bottle for solids
    25 g
    Log in for availability
    Log in for availability
    1 * 25 g
    1 * 25 g - 4.150,00 kr
    4.150,00 kr
    1 * 25 g
       4.150,00 kr

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.