4,4'-Methylenedi-o-toluidine, Supelco®
Catalog # 46106-100MG
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:100 mg
- Seq:2440
Specifications
About this item
:
:226.3200 g/mol
:M-Clarity™ quality level = MQ100
EINECS:212-658-8
CAS Number:838-88-0
Formula:C₁₅H₁₈N₂
MDL Number:MFCD00126963