N(α)-Cbz-DL-tryptophan ≥97.0% (by titrimetric analysis)
Supplier: TCI
Certificates
About this item
CAS Number:13058-16-7
:
Formula:C₁₉H₁₈N₂O₄
MDL Number:MFCD00069707
:338.4 g/mol
EINECS:235-949-1
Specification Test Results
- Appearance:White to Yellow to Orange powder to crystal
- Purity (Neutralization titration):min. 97.0 %
- Melting point:166.0 to 171.0 deg-C
- Solubility in Methanol:almost transparency
Specification Test Results
Frequently Bought Together