4,4'-Methylenebis(2-chloroaniline) ≥90.0% (by HPLC)
Supplier: TCI
Certificates
About this item
CAS Number:101-14-4
:
Formula:C₁₃H₁₂Cl₂N₂
MDL Number:MFCD00047829
Merck Index:14,06059
EINECS:202-918-9
UN:3077
ADR:9,III
:GHS07
:GHS08
:GHS09
Specification Test Results
- Appearance:Light orange to Yellow to Green powder to crystal
- Purity (HPLC):min. 90.0 area%
- Purity (Nonaqueous Titration):min. 98.0 %
- Melting point:104.0 to 109.0 deg-C
Specification Test Results
Frequently Bought Together