Order Entry
Denmark
ContactUsLinkComponent
4,4'-Methylenebis(2-chloroaniline)
4,4'-Methylenebis(2-chloroaniline)
Catalog # EHERC15087500
Supplier:  EHRENSTORFER
CAS Number:  
4,4'-Methylenebis(2-chloroaniline)
Catalog # EHERC15087500
Supplier:  EHRENSTORFER
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    0,1 g
  • Seq:
    3020

Specifications

About this item

 :
EINECS:202-918-9
CAS Number:101-14-4
Formula:C₁₃H₁₂Cl₂N₂
MDL Number:MFCD00047829