N(α)-Boc-L-tryptophan ≥98.0% (by HPLC)
TCIAB1639-25G
: TCI
:
- Pack type:Glass bottle
- Pk:25 g
- Seq:3308
- Environmentally Preferable:
- Appearance:White to Light yellow to Light orange powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (Neutralization titration):min. 97.0 %
- Specific rotation [a]20/D:-18.0 to -22.0 deg (C=2, DMF)
: Boc-Trp-OH , Boc-Trp-OH
:
:304.3 g/mol
:236-072-7
:13139-14-5
:C₁₆H₂₀N₂O₄
:MFCD00065595